Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mepyramine maleate, Tocris Bioscience™
SDP

Catalog No. 0660100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-601-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-601-00 Supplier R&D Systems Supplier No. 0660/100

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective H1 inverse agonist

Selective inverse agonist for the H1 receptor. Inhibits histamine induced inositol phosphate (InsP) production (log EC50 = -7.94) and intracellular calcium mobilization. Sequesters Gq/11 protein, reducing its availability for other receptors associated with the same signaling pathway.

Chemical Identifiers

CAS 59-33-6
Molecular Formula C21H27N3O5
Molecular Weight (g/mol) 401.46
MDL Number MFCD00069333
InChI Key JXYWFNAQESKDNC-BTJKTKAUSA-N
Synonym pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
PubChem CID 5284451
IUPAC Name (2Z)-but-2-enedioic acid; N-[2-(dimethylamino)ethyl]-N-[(4-methoxyphenyl)methyl]pyridin-2-amine
SMILES OC(=O)\C=C/C(O)=O.COC1=CC=C(CN(CCN(C)C)C2=CC=CC=N2)C=C1

Specifications

Quantity 100 mg
Formula Weight 401.46
Percent Purity >98%
Chemical Name or Material Mepyramine maleate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.