Learn More
Cayman Chemical Mepyramine (maleate), Purity: ≥95%, CAS: 59-33-6, 5 g
SDP
Catalog No. 502984986
Encompass_Preferred
A histamine H1 receptor inverse agonist that inhibits histamine-induced inositol phosphate production with a log EC50 value of -7.94
Catalog No. 50-298-4986 Supplier Cayman Chemical Supplier No. 209785 G
A histamine H1 receptor inverse agonist that inhibits histamine-induced inositol phosphate production with a log EC50 value of -7.94