Learn More
Apexbio Technology LLC Mequinol 150-76-5 50mg
SDP
Catalog No. 502240697
Encompass_Preferred
Mequinol (150-76-5) is a small-molecule inhibitor targeting tyrosinase It is designed to inhibit tyrosinase activity thereby modulating melanogenesis pathways related to skin pigmentation Mequinol exerts its biological activity primarily through inhibition of tyrosinase and displays antioxidant properties cooperating with other phenolic antioxidants to intervene in oxidative mechanisms potentially related to carcinogenesis In addition Mequinol activates human estrogen receptor (hER )-dependent molecular chaperone proteins indicating estrogen-like activities Based on these pharmacological properties Mequinol holds research potential in studies on melanin biosynthesis regulation anti-oxidative cellular responses and estrogen signaling pathway exploration
This item is not returnable. View return policy
Catalog No. 50-224-0697 Supplier Apexbio Technology LLC Supplier No. B196850
Mequinol (150-76-5) is a small-molecule inhibitor targeting tyrosinase It is designed to inhibit tyrosinase activity thereby modulating melanogenesis pathways related to skin pigmentation Mequinol exerts its biological activity primarily through inhibition of tyrosinase and displays antioxidant properties cooperating with other phenolic antioxidants to intervene in oxidative mechanisms potentially related to carcinogenesis In addition Mequinol activates human estrogen receptor (hER )-dependent molecular chaperone proteins indicating estrogen-like activities Based on these pharmacological properties Mequinol holds research potential in studies on melanin biosynthesis regulation anti-oxidative cellular responses and estrogen signaling pathway exploration