Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mequinol-d4 | 126840-02-6 | 98.9% | 128.16 | 5 MG
SDP

Catalog No. 5000400475
Encompass_Preferred

Mequinol-d4 is a laboratory chemical primarily used for research. It is presented as a solid and is stable under recommended storage conditions. This substance is not classified as hazardous and is intended for experienced personnel in appropriately equipped facilities.

  • Used as a laboratory chemical
  • Not classified as a hazardous substance
  • Stable under recommended storage conditions
  • Available as a solid
  • Recommended storage for powder: -20°C for 3 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • High purity (98.9%)

Catalog No. 50-004-00475 Supplier Medchemexpress LLC Supplier No. HY30270S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mequinol-d4 is a laboratory chemical primarily used for research. It is presented as a solid and is stable under recommended storage conditions. This substance is not classified as hazardous and is intended for experienced personnel in appropriately equipped facilities.

  • Used as a laboratory chemical
  • Not classified as a hazardous substance
  • Stable under recommended storage conditions
  • Available as a solid
  • Recommended storage for powder: -20°C for 3 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • High purity (98.9%)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.