Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mequinol-d4 | 126840-02-6 | 98.9% | 128.16 | 50 MG
SDP

Catalog No. 5000400620
Encompass_Preferred

Mequinol-d4 is a chemical substance primarily intended for laboratory research and the manufacture of other substances. This product is for research use only and should be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities. It presents as a white to off-white solid.

  • Intended for laboratory chemical applications
  • Stable under recommended storage conditions
  • Can be stored as powder at -20°C for 3 years
  • Can be stored in solvent at -80°C for 6 months
  • Appears as a white to off-white solid

Catalog No. 50-004-00620 Supplier Medchemexpress LLC Supplier No. HY30270S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mequinol-d4 is a chemical substance primarily intended for laboratory research and the manufacture of other substances. This product is for research use only and should be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities. It presents as a white to off-white solid.

  • Intended for laboratory chemical applications
  • Stable under recommended storage conditions
  • Can be stored as powder at -20°C for 3 years
  • Can be stored in solvent at -80°C for 6 months
  • Appears as a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.