Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mequinol-d4 (4-methoxyphenol-d4) | 126840-02-6 | 128.16 | C7H4D4O2 | 10 MG
SDP

Catalog No. 5000297581
Encompass_Preferred

Mequinol-d4 is the d4 isotopologue of 4-methoxyphenol supplied as a 10 mg powder for research use. It is intended as a stable isotope reference standard and analytical reagent for tracer studies, method development, and mass spectrometry applications. For research use only; not for human or clinical use.

  • Deuterium-labeled reference standard for analytical applications.
  • Suitable as an internal standard for mass spectrometry and tracer studies.
  • Supplied as a 10 mg powder for laboratory use.
  • Molecular weight 128.16; chemical formula C7H4D4O2.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-97581 Supplier Medchemexpress LLC Supplier No. HY30270S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mequinol-d4 is the d4 isotopologue of 4-methoxyphenol supplied as a 10 mg powder for research use. It is intended as a stable isotope reference standard and analytical reagent for tracer studies, method development, and mass spectrometry applications. For research use only; not for human or clinical use.

  • Deuterium-labeled reference standard for analytical applications.
  • Suitable as an internal standard for mass spectrometry and tracer studies.
  • Supplied as a 10 mg powder for laboratory use.
  • Molecular weight 128.16; chemical formula C7H4D4O2.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.