Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mequitazine | 29216-28-2 | 322.47 g/mol | C20H22N2S | 10 MG
SDP

Catalog No. 5000297650
Encompass_Preferred

An analytical reference standard of the antihistamine mequitazine supplied as a powdered solid for preparation of reference solutions in research and analytical workflows. Intended for laboratory use and not validated for medical or therapeutic applications.

  • Analytical standard for research and analytical applications.
  • Powder form suitable for dissolution and stock solution preparation.
  • CAS number 29216-28-2 for unambiguous identification.
  • Molecular formula C20H22N2S and molecular weight 322.47 g/mol.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years) as recommended for stability.

Catalog No. 50-002-97650 Supplier Medchemexpress LLC Supplier No. HYB2168R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An analytical reference standard of the antihistamine mequitazine supplied as a powdered solid for preparation of reference solutions in research and analytical workflows. Intended for laboratory use and not validated for medical or therapeutic applications.

  • Analytical standard for research and analytical applications.
  • Powder form suitable for dissolution and stock solution preparation.
  • CAS number 29216-28-2 for unambiguous identification.
  • Molecular formula C20H22N2S and molecular weight 322.47 g/mol.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years) as recommended for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.