Learn More
Eurofins Discoverx Mer, active, 10 µg. (14-728)
Supplier:  Eurofins Discoverx 14728
Encompass
N-terminal GST-tagged recombinant, human Mer amino acids 557-882. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9245