Learn More
Carnabio Usa Inc MER(MERTK)/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 081085
Encompass
MER(MERTK), Cytoplasmic domain, Wild type, Amino Acid 528-999, NP_006334.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0513