Learn More
Cayman Chemical MerafloxacIn 10mg
SDP
Catalog No. 502916846
Encompass_Preferred
A fluoroquinolone antibiotic; active against a variety of Gram-positive (MIC50s = 0.12-0.5 UG/ml) and Gram-negative bacteria (MIC50s = 0.06-1 UG/ml)
Catalog No. 50-291-6846 Supplier Cayman Chemical Supplier No. 3770910 MG
A fluoroquinolone antibiotic; active against a variety of Gram-positive (MIC50s = 0.12-0.5 UG/ml) and Gram-negative bacteria (MIC50s = 0.06-1 UG/ml)