Learn More
Medchemexpress LLC HY-139010 10mg , Merafloxacin CAS:91188-00-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13901010MG
Encompass_Preferred
HY-139010 10mg Merafloxacin CAS: 91188-00-0 Merafloxacin (CI-934), a fluoroquinolone antibacterial agent, is a selective programmed -1 ribosomal frameshifting (-1 PRF) inhibitor of beta coronaviruses. Merafloxacin exhibits in vitro activity against gram-positive and gram-negative bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1436