Learn More
AdipoGen Merafloxacin
SDP
Catalog No. 502562170
Encompass_Preferred
Chemical. CAS 91188-00-0. Formula C19H23F2N3O3. MW 379.4. Merafloxacin is a fluoroquinolone broad-spectrum antibacterial compound. It is an inhibitor of bacterial DNA gyrase and Type II DNA topoisomerase. It demonstrated excellent activity against Gram-positive organisms and less potent activity against Gram-negative bacteria. Merafloxacin is a programmed -1 ribosomal frameshifting -1 PRF inhibitor of SARS-CoV-2. Frameshift inhibition by merafloxacin is robust to mutations within the pseudoknot region and is similarly effective on -1 PRF of other betacoronaviruses and blocks SARS-CoV-2 replication in Vero E6 cells. The compound reduced viral levels in infected African green monkey VeroE6 cells in a concentration-dependent manner. Merafloxacin showed no cellular toxicity and resulted in a 3 to 4 orders of magnitude reduction of SARS-CoV-2 titer, with IC50 of 4.3 µMu.
Catalog No. 50-256-2170 Supplier AdipoGen Supplier No. AGCR13756M005
Chemical. CAS 91188-00-0. Formula C19H23F2N3O3. MW 379.4. Merafloxacin is a fluoroquinolone broad-spectrum antibacterial compound. It is an inhibitor of bacterial DNA gyrase and Type II DNA topoisomerase. It demonstrated excellent activity against Gram-positive organisms and less potent activity against Gram-negative bacteria. Merafloxacin is a programmed -1 ribosomal frameshifting -1 PRF inhibitor of SARS-CoV-2. Frameshift inhibition by merafloxacin is robust to mutations within the pseudoknot region and is similarly effective on -1 PRF of other betacoronaviruses and blocks SARS-CoV-2 replication in Vero E6 cells. The compound reduced viral levels in infected African green monkey VeroE6 cells in a concentration-dependent manner. Merafloxacin showed no cellular toxicity and resulted in a 3 to 4 orders of magnitude reduction of SARS-CoV-2 titer, with IC50 of 4.3 µMu.