Learn More
TARGETMOL CHEMICALS INC MERAFLOXACIN 5MG
SDP
Catalog No. 502576213
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Merafloxacin (CI 934) is a fluoroquinolone antibacterial which was also identified as a 1 PRF inhibitor of SARS-CoV-2. purity: 98%
Catalog No. 50-257-6213 Supplier TARGETMOL CHEMICALS INC Supplier No. T90665MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Merafloxacin (CI 934) is a fluoroquinolone antibacterial which was also identified as a 1 PRF inhibitor of SARS-CoV-2. purity: 98%