Learn More
Martor Secupro Merak
Catalog No. NC0348589
Encompass_Preferred
The Merak is an ergonomically designed safety knife that is a great combination of safety with ease of use. The operator exposes the blade by simply squeezing the trigger and the blade automatically retracts at the end of the cut.-Fully automatic retractable blade for safety-Ergonomically friendly trigger mechanism to engage the blade-Durable body made of polycarbonate plastic-Safer alternative to fixed blade utility knives for general cutting tasks-1/2" of blade extension
Catalog No. NC0348589 Supplier Martor Supplier No. 124001.02
The Merak is an ergonomically designed safety knife that is a great combination of safety with ease of use. The operator exposes the blade by simply squeezing the trigger and the blade automatically retracts at the end of the cut.-Fully automatic retractable blade for safety-Ergonomically friendly trigger mechanism to engage the blade-Durable body made of polycarbonate plastic-Safer alternative to fixed blade utility knives for general cutting tasks-1/2" of blade extension