Learn More
AdipoGen Meranzin hydrate
SDP
Catalog No. 502560462
Encompass_Preferred
Chemical. CAS 5875-49-0. Formula C15H18O5. MW 278.3. Meranzin is a bioactive compound from the Traditional Chinese Medicine TCM Chaihu-Shugan-San CSS. Meranzin exhibits anti-inflammatory, analgesic and antibacterial activity. Meranzin regulates the alpha 2-adrenoceptor and involves the AMPA-ERK1/2–BDNF signaling pathway. Meranzin has the potential for the prevention of the comorbidity of atherosclerosis and depression.
Catalog No. 50-256-0462 Supplier AdipoGen Supplier No. CDXM0513M001
Chemical. CAS 5875-49-0. Formula C15H18O5. MW 278.3. Meranzin is a bioactive compound from the Traditional Chinese Medicine TCM Chaihu-Shugan-San CSS. Meranzin exhibits anti-inflammatory, analgesic and antibacterial activity. Meranzin regulates the alpha 2-adrenoceptor and involves the AMPA-ERK1/2–BDNF signaling pathway. Meranzin has the potential for the prevention of the comorbidity of atherosclerosis and depression.