Learn More
Medchemexpress LLC HY-19024 5mg Medchemexpress, Merbarone CAS:97534-21-9 Purity:>98%
SDP
Catalog No. 502082107
Encompass_Preferred
Medchemexpress, HY-19024 5mg Merbarone CAS:97534-21-9 Merbarone (NSC 336628) is an orally active inhibitor of topoisomerase II. Merbarone acts primarily by blocking topoisomerase II-mediated DNA cleavage without stabilizing topo II-DNA covalent complexes. Merbarone is an anticancer agent[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2107 Supplier Medchemexpress LLC Supplier No. HY190245MG
Medchemexpress, HY-19024 5mg Merbarone CAS:97534-21-9 Merbarone (NSC 336628) is an orally active inhibitor of topoisomerase II. Merbarone acts primarily by blocking topoisomerase II-mediated DNA cleavage without stabilizing topo II-DNA covalent complexes. Merbarone is an anticancer agent[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.