Learn More
Medchemexpress LLC Merbromin | 129-16-8 | 500 MG
SDP
Catalog No. 5000408556
Encompass_Preferred
Merbromin is a mixed inhibitor of the 3-chymotrypsin-like protease (3CLpro) of SARS-CoV-2, with an IC50 of 2.7 μM. It effectively inhibits the replication of the Zika virus (ZIKV) and shows anti-ZIKV activity by inhibiting ZIKVpro. Merbromin can also be used as a topical disinfectant for cuts and scrapes, and it serves as a biological dye.
  • Mixed inhibitor of 3-chymotrypsin-like protease (3CLpro) of SARS-CoV-2.
  • Effectively inhibits replication of the Zika virus (ZIKV).
  • Shows anti-ZIKV activity by inhibiting ZIKVpro.
  • Can be used as a topical disinfectant for cuts and scrapes.
  • Functions as a biological dye.
Catalog No. 50-004-08556 Supplier Medchemexpress LLC Supplier No. HYB0961500MG
Merbromin is a mixed inhibitor of the 3-chymotrypsin-like protease (3CLpro) of SARS-CoV-2, with an IC50 of 2.7 μM. It effectively inhibits the replication of the Zika virus (ZIKV) and shows anti-ZIKV activity by inhibiting ZIKVpro. Merbromin can also be used as a topical disinfectant for cuts and scrapes, and it serves as a biological dye.
  • Mixed inhibitor of 3-chymotrypsin-like protease (3CLpro) of SARS-CoV-2.
  • Effectively inhibits replication of the Zika virus (ZIKV).
  • Shows anti-ZIKV activity by inhibiting ZIKVpro.
  • Can be used as a topical disinfectant for cuts and scrapes.
  • Functions as a biological dye.