Learn More
Gelest Inc Mercaptopropyl terminated polydimethylsiloxane, 80-120 cSt | 308072-58-4 | 100g
Catalog No. NC1391064
Encompass
This item has a minimum qty of 2 per supplier requirements.
Mercaptopropyl terminated polydimethylsiloxane, 80-120 cSt | MW: 10000 | 308072-58-4 | 100g
Catalog No. NC1391064 Supplier Gelest Inc Supplier No. DMSSM21100G
This item has a minimum qty of 2 per supplier requirements.
Mercaptopropyl terminated polydimethylsiloxane, 80-120 cSt | MW: 10000 | 308072-58-4 | 100g