Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number AC148890010
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
367-51-1
C2H3NaO2S
114.09
MFCD00043386
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
23690444
CHEBI:86481
sodium;2-sulfanylacetate
C(C(=O)[O-])S.[Na+]
367-51-1
C2H3NaO2S
114.09
MFCD00043386
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
23690444
CHEBI:86481
sodium;2-sulfanylacetate
C(C(=O)[O-])S.[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CO2Na
Quantity 1 kg
Merck Index 15,8819
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 114.09
Percent Purity 98%
Chemical Name or Material Mercaptoacetic acid, sodium salt, 98%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin
EINECSNumber 206-696-4
RTECSNumber AI7700000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit