Learn More
Strem, An Ascensus Company CAS# 112926-00-8. 10g. Metals scavenging agent, Mercaptopropyl modified silica (BASF MSA-FC Si-3). MFCD07370733
SDP
Catalog No. NC1769966
Encompass
CAS# 112926-00-8. 10g. Metals scavenging agent, Mercaptopropyl modified silica (BASF MSA-FC Si-3). MFCD07370733. Molecular Weight: . Molecular Formula: . Color/form: d50=83mum; white to slightly yellow pwdr. Strem# 14-4353. http://www.strem.com/catalog/v/14-4353/
Catalog No. NC1769966 Supplier Strem, An Ascensus Company Supplier No. 14435310G
CAS# 112926-00-8. 10g. Metals scavenging agent, Mercaptopropyl modified silica (BASF MSA-FC Si-3). MFCD07370733. Molecular Weight: . Molecular Formula: . Color/form: d50=83mum; white to slightly yellow pwdr. Strem# 14-4353. http://www.strem.com/catalog/v/14-4353/