Learn More
Apexbio Technology LLC Mercaptopurine (6-MP) 50-44-2 25mg
SDP
Catalog No. 5000096638
Encompass_Preferred
Mercaptopurine (6-MP CAS 50-44-2) is a purine analog that disrupts de novo purine biosynthesis by incorporating into nucleic acids thereby inhibiting DNA and RNA synthesis This mode of action results in cytotoxic and immunosuppressive effects making the compound a valuable tool in studies of cell proliferation immune response modulation and leukemia research 6-MP serves as the active metabolite of azathioprine and it is also used to investigate the interaction between purine metabolism inhibitors and anticoagulant agents such as warfarin
Catalog No. 50-000-96638 Supplier Apexbio Technology LLC Supplier No. A235525
Mercaptopurine (6-MP CAS 50-44-2) is a purine analog that disrupts de novo purine biosynthesis by incorporating into nucleic acids thereby inhibiting DNA and RNA synthesis This mode of action results in cytotoxic and immunosuppressive effects making the compound a valuable tool in studies of cell proliferation immune response modulation and leukemia research 6-MP serves as the active metabolite of azathioprine and it is also used to investigate the interaction between purine metabolism inhibitors and anticoagulant agents such as warfarin