Learn More
Selleck Chemical LLC Mercaptopurine 6-MP-50mg
SDP
Catalog No. 502389687
Encompass_Preferred
Mercaptopurine(6-MP) is a widely used antileukemic agent and immunosuppressive drug that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA.
Catalog No. 50-238-9687 Supplier Selleck Chemical LLC Supplier No. S130550MG
Mercaptopurine(6-MP) is a widely used antileukemic agent and immunosuppressive drug that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA.