Learn More
Selleck Chemical LLC Mercaptopurine (6-MP) S1305-10mM/1mL
SDP
Catalog No. 503108256
Encompass_Preferred
Mercaptopurine(6-MP) is a widely used antileukemic agent and immunosuppressive drug that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA
Catalog No. 50-310-8256 Supplier Selleck Chemical LLC Supplier No. S130510MM/1ML
Mercaptopurine(6-MP) is a widely used antileukemic agent and immunosuppressive drug that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA