Learn More
Selleck Chemical LLC Mercaptopurine (6-MP) S1305-1g
SDP
Catalog No. 503110989
Encompass_Preferred
Mercaptopurine(6-MP) is a widely used antileukemic agent and immunosuppressive drug that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA
Catalog No. 50-311-0989 Supplier Selleck Chemical LLC Supplier No. S13051G
Mercaptopurine(6-MP) is a widely used antileukemic agent and immunosuppressive drug that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA