Learn More
Mercedes Medical Glass Coverslips
SDP
Catalog No. 1518384
Change view
Click to view available options
Quantity:
330/box, 10 boxes/case
Catalog No. Quantity
15-183-84 330/box, 10 boxes/case
1 options
This item is not returnable. View return policy
Catalog No. 15-183-84 Supplier Mercedes Medical Supplier No. MER R0012
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Product Type Coverslip
Shape Circle
Material Borosilicate Glass
Thickness (Metric) #1 (0.13 to 0.16 mm)
Diameter (Metric) 12 mm
For Use With (Equipment) Leica™ recommended
Quantity 330/box, 10 boxes/case