Learn More
Med Vet International Merck Estrumate, 10 Dose Vial
Catalog No. 502827937
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Estrumate is used as a luteolytic agent in cattle. The luteolytic action of Estrumate can be utilized to manipulate the estrous cycle to better fit certain management practices, terminal pregnancies resulting from mismatings, and to treat certain conditions associated with prolonged luteal function.
Catalog No. 50-282-7937 Supplier Med Vet International Supplier No. RXESTRUMATE10D
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Estrumate is used as a luteolytic agent in cattle. The luteolytic action of Estrumate can be utilized to manipulate the estrous cycle to better fit certain management practices, terminal pregnancies resulting from mismatings, and to treat certain conditions associated with prolonged luteal function.