Learn More
Med Vet International Otomax Ointment, 30mL, 6/Pack
Catalog No. 502828478
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Combination steroid, antibacterial, and antifungal ointment used to treat acute and chronic otitis. Active ingredients are Betamethasone Valerate, Gentamicin Sulfate and Clotrimazole. For otic use in dogs only
Catalog No. 50-282-8478 Supplier Med Vet International Supplier No. RXOTOMAX30
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Combination steroid, antibacterial, and antifungal ointment used to treat acute and chronic otitis. Active ingredients are Betamethasone Valerate, Gentamicin Sulfate and Clotrimazole. For otic use in dogs only