Learn More
ATCC Merck Pneumococcal polysaccharide powder Type 12 (d12F) 2mg
Catalog No. 502383579
Encompass
This purified pneumococcal polysaccharide was manufactured by Merck & Company for small-quantity distribution by ATCC. The pneumococcal powders include types that are components of the current 23-valent vaccine against Streptococcus pneumoniae. Pneumococcal Polysaccharide Powder Type 12F (US Type 12) antigens can be used in the polyvalent vaccine are extracted and purified separately. These vaccine products can be used to assay for type-specific antibodies, research immune responses, and immunodeficiencies.
Catalog No. 50-238-3579 Supplier ATCC Supplier No. 22X
This purified pneumococcal polysaccharide was manufactured by Merck & Company for small-quantity distribution by ATCC. The pneumococcal powders include types that are components of the current 23-valent vaccine against Streptococcus pneumoniae. Pneumococcal Polysaccharide Powder Type 12F (US Type 12) antigens can be used in the polyvalent vaccine are extracted and purified separately. These vaccine products can be used to assay for type-specific antibodies, research immune responses, and immunodeficiencies.