Learn More
ATCC Merck Pneumococcal polysaccharide powder Type 2 (d2) 200mg
Catalog No. 502383073
Encompass
Purified pneumococcal polysaccharides are manufactured by Merck & Company for small quantity distribution by ATCC. The pneumococcal powders include those types which are components of the current 23-valent vaccine against Streptococcus pneumoniae.Each of the polysaccharide antigens used in the polyvalent vaccine is extracted and purified separately. These vaccine products can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.
Catalog No. 50-238-3073 Supplier ATCC Supplier No. 168X
Purified pneumococcal polysaccharides are manufactured by Merck & Company for small quantity distribution by ATCC. The pneumococcal powders include those types which are components of the current 23-valent vaccine against Streptococcus pneumoniae.Each of the polysaccharide antigens used in the polyvalent vaccine is extracted and purified separately. These vaccine products can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.