Learn More
ATCC Merck Pneumococcal polysaccharide powder Type 23 (d23F) 2mg
Catalog No. 502382584
Encompass
This purified pneumococcal polysaccharides was manufactured by Merck & Company for small-quantity distribution by ATCC. The pneumococcal powders include types that are components of the current 23-valent vaccine against Streptococcus pneumoniae. Each of the polysaccharide antigens used in the polyvalent vaccine is extracted and purified separately. These vaccine products can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2584 Supplier ATCC Supplier No. 25X
This purified pneumococcal polysaccharides was manufactured by Merck & Company for small-quantity distribution by ATCC. The pneumococcal powders include types that are components of the current 23-valent vaccine against Streptococcus pneumoniae. Each of the polysaccharide antigens used in the polyvalent vaccine is extracted and purified separately. These vaccine products can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.