Learn More
Mercodia Inc MERCODIA LP(A) ELISA
SDP
Supplier:  Mercodia Inc 10110601
Encompass_Preferred
Immunoassay for quantitative determination of human lipoprotein(a) in serum or plasma. Easy-to-use assay Very low lot-to-lot variation Short incubation time Calibrated against a highly purified, fully validated, commercial Lp(a) preparation.
Catalog No. NC1253320