Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric acetate, 99+%

Catalog No. p-44660
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC255650050 5 g Glass bottle
AC255651000 100 g Glass bottle
AC255650010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. AC255650050 Supplier Thermo Scientific Chemicals Supplier No. 255650050
Only null left
Add to Cart
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 1600-27-7 | C4H6HgO4 | 318.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.67
MDL Number MFCD00012165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Melting Point 179.0°C to 182.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula (CH3CO2)2Hg
Beilstein 02, 96
Fieser 01,644; 02,264; 03,194; 04,319; 05,424; 06,358; 10,252; 11,315; 12,298; 17,176
Merck Index 15, 5940
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 318.67
Percent Purity ≥99%
Infrared Spectrum Authentic
Packaging Glass bottle
Residue after Ignition 0.02% max. (substances not reducible with formic acid)
Chemical Name or Material Mercuric acetate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Fatal in contact with skin.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-491-1
RTECSNumber AI8575000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.