Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric acetate, ACS reagent

Catalog No. AC447271000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447271000 100 g Plastic bottle
AC447270250 25 g Glass Bottle
AC447275000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC447271000 Supplier Thermo Scientific Chemicals Supplier No. 447271000
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1600-27-7 | C4H6HgO4 | 318.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.67
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Melting Point 179.0°C to 182.0°C
Quantity 100 g
Linear Formula (CH3CO2)2Hg
Merck Index 15,594
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 318.67
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Mercuric acetate
Hazard Category GHS Signal Word: Danger
EINECSNumber 216-491-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.