Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Chloride, 5% (w/v), LabChem™

Catalog No. LC166002 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC166002 1 L
LC166001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC166002 Supplier LabChem, Inc. Supplier No. LC166002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7487-94-7 , 7732-18-5
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name mercury(2+) dichloride
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Color Colorless
CAS Max % 95, 5
Physical Form Liquid
Quantity 1 L
Linear Formula HgCl2
UN Number UN2024
Solubility Information Soluble in water
Formula Weight 271.5
Percent Purity 5% (w/v)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Mercury
Chemical Name or Material Mercuric Chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (nervous system) through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off immediately all contaminated clothing and wash it before reuse.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.