Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Iodide, Red, Powder, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611563 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
1 kg
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Drum
Fiber Drum
Poly Bottle
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-563 25 g Amber Glass Bottle
18-611-564 125 g Amber Glass Bottle
18-611-565 500 g Amber Glass Bottle
18-611-566 1 kg Poly Bottle
18-611-567 2.5 kg Amber Glass Bottle
18-611-568 12 kg Poly Pail
18-611-569 25 kg Fiber Drum
18-611-570 50 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-611-563 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M116025GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.40
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
IUPAC Name mercury(2+) diiodide
SMILES [I-].[I-].[Hg++]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.