Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Nitrate, 0.141 N, 0.0705 M, 1 mg = 5 mL Cl-, Ricca Chemical
SDP

Catalog No. p-2916703 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4740-16 500 mL
4740-32 1 L
4740-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. 4740-16 Supplier Ricca Chemical Company Supplier No. 474016
Only null left
Add to Cart
Add to Cart
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

APHA/EPA for Chloride Testing

Specifications

Volume (Metric) 500 mL
Molecular Formula H2HgN2O7
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Packaging Amber Glass Bottle
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name mercury(2+) hydrate dinitrate
Molecular Weight (g/mol) 342.62
Quantity 500 mL
Test Method APHA/EPA for Chloride
Type Mercuric Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.