Learn More
Hach Company Mercuric Nitrate Digital Titrator Cartridge, 0.2256 N
Catalog No. 501995017 Shop All Hach Company Products
Encompass
For chloride determination by Digital Titrator titration with mercuric nitrate, using diphenylcarbazone indicator. Hach method 8206. Digits = mg/L Cl- in 100 mL sample. Approximately 40-100 tests
Catalog No. 50-199-5017 Supplier Hach Company Supplier No. 1439301
For chloride determination by Digital Titrator titration with mercuric nitrate, using diphenylcarbazone indicator. Hach method 8206. Digits = mg/L Cl- in 100 mL sample. Approximately 40-100 tests