Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Nitrate, (1mL = 0.5mg Cl-), for Chloride, Certified, 0.0141N ±0.0002N (0.0071M), LabChem™

Catalog No. LC166501 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC166501 500 mL
LC166502 1 L
LC166504 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC166501 Supplier LabChem, Inc. Supplier No. LC166501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22852-67-1 , 7732-18-5 , 7697-37-2
PubChem CID 91886652  

Specifications

Color Colorless
CAS Max % 0.25, 99.65, 0.1
Linear Formula Hg(NO3)2·H2O
Concentration 0.0141N ±0.0002N (0.0071M)
Solubility Information Soluble in water
Formula Weight 342.62
Grade Certified
Identification Passes Test
Packaging Amber Glass
Traceability to NIST Traceable to NIST
Decomposition Information Nitrogen oxides; Mercury
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Mercuric Nitrate, (1mL = 0.5mg Cl-), for Chloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.