Learn More
Mercuric Nitrate, (1mL = 1mg NaCl), Certified, 0.0171N ±0.0002N (0.0086M), LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| CAS Max % | 0.1, 99.59, 0.31 |
| Quantity | 4 L |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Specific Gravity | 1g/mL |
| Decomposition Information | Nitrogen oxides; Mercury |
| Density | 1g/mL |
| Linear Formula | Hg(NO3)2·H2O |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful in contact with skin. May cause damage to organs (nervous system, liver, kidneys) through prolonged or repeated exposure. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves, protective clothing, eye protection. Avoid release to the environment. If on skin: Wash with plenty of soap and water. Call a poison center/doctor if you feel unwell. Take off contaminated clothing and wash it before reuse. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.