Learn More
Hach Company Mercuric Nitrate Digital Titrator Cartridge, 2.256 N
Catalog No. 501994466 Shop All Hach Company Products
Encompass
For chloride determination by mercuric nitrate titration. Hach Method 8206. Digits x 1.0 = mg/L Cl- in 100 mL sample. Approximately 40-100 tests.
Catalog No. 50-199-4466 Supplier Hach Company Supplier No. 92101
For chloride determination by mercuric nitrate titration. Hach Method 8206. Digits x 1.0 = mg/L Cl- in 100 mL sample. Approximately 40-100 tests.