Learn More
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 0.5, 97.7, 1.8 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Linear Formula | Hg(NO3)2·H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 342.62 |
| Grade | Certified |
| Concentration or Composition (by Analyte or Components) | 0.100N ±0.001N (0.05M) |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed or if inhaled. May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Do not eat, drink or smoke when using this product. Wash exposed skin after handling. Use only outdoors or in a well-ventilated area. Avoid release to the environment. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. Collect spillage. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.