Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Nitrate, For Chloride, Certified, 0.0250N ±0.0005N (0.013M), LabChem™

Catalog No. LC166571
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC166571 500 mL
LC166572 1 L
LC166574 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC166571 Supplier LabChem, Inc. Supplier No. LC166571
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5 , 22852-67-1
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
CAS Max % 0.2, 99.35, 0.45
Linear Formula Hg(NO3)2·H2O
Concentration 0.0250N ±0.0005N (0.013M)
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 342.62
Grade Certified
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Nitrogen oxides; Mercury
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Mercuric Nitrate, For Chloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.