Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Sulfate, ACS Reagent (Mercury (II) Sulfate), Reagents

Catalog No. 01337893
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-893 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-893 Supplier Reagents Holdings Llc Supplier No. C1248000100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mercuric Sulfate, ACS Reagent (Mercury (II) Sulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym Mercury(II) Sulfate
IUPAC Name mercury(2+) sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 450°C
Color White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Appearance White
Specific Gravity 6.47
Chemical Name or Material Mercuric Sulfate, ACS Reagent (Mercury (II) Sulfate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.