Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Thiocyanate, Reagent Grade, LabChem™

Catalog No. LC167108 Shop All LabChem Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC167108 25 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC167108 Supplier LabChem, Inc. Supplier No. LC167108
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 592-85-8
Molecular Formula C4HgK2N4S4
Molecular Weight (g/mol) 511.10
InChI Key HWXOCCNUUYRBKP-UHFFFAOYSA-J
IUPAC Name mercury(2+) dipotassium tetrakis(cyanosulfanide)
SMILES [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N

Specifications

Melting Point 165°C
Color White
Physical Form Crystals
CAS Max % 100
Quantity 25 g
Identification Passes Test
Packaging Amber Glass
Specific Gravity 3.72g/cm3
Decomposition Information Hydrogen cyanide; Mercury
Density 3.72g/cm3
Linear Formula Hg(CHNS)2
UN Number UN1646
Solubility Information Soluble in water
Formula Weight 316.75
Grade Reagent
Chemical Name or Material Mercuric Thiocyanate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Fatal if swallowed or if inhaled.
Toxic in contact with skin.
May cause damage to organs (nervous system) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe dust.
Wear eye protection, protective gloves, protective clothing.
Wear respiratory protection.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Immediately call a poison center/doctor.
Rinse mouth.
If on skin: Wash with plenty of soap and water.
Immediately call a poison center/doctor.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
Remove/Take off immediately all contaminated clothing and wash before reuse.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.