Learn More
Hach Company Mercuric Thiocyanate Solution, 200 mL
Supplier:  Hach Company 2212129
Encompass
For colorimetric chloride determination by the mercuric thiocyanate method. Hach method 8113. Range: 0.1 to 25.0 mg/L Cl- 200 mL bottle
Catalog No. 50-199-5560