Learn More
NE MICROSCOPE COMPANY MERCURY BURNER (HBO103W/2)
Catalog No. NC9160986
Encompass_Preferred
Lamp; Burner; Mercury; Short Arc; Osram specialty bulb; HBO103W/2; Used in Scientific equipment including Microscopes; 100W; Current: DC; 23V; Operating Current: 4.3; Luminous Flux: 3000; Lamp Life: 300 hr.; 90L x 10mm D; Contains mercury
Catalog No. NC9160986 Supplier NE MICROSCOPE COMPANY Supplier No. HUB257
Lamp; Burner; Mercury; Short Arc; Osram specialty bulb; HBO103W/2; Used in Scientific equipment including Microscopes; 100W; Current: DC; 23V; Operating Current: 4.3; Luminous Flux: 3000; Lamp Life: 300 hr.; 90L x 10mm D; Contains mercury