Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury (Certified ACS), Fisher Chemical

Catalog No. M1411LB Shop All Fisherbrand Products
Change view
Click to view available options
Poly Bottle; 1 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
M141-1LB Poly Bottle; 1 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M141-1LB

missing translation for 'validMainframeMsg'

Specifications

Color Silver
Melting Point -38.87°C
CAS 7439-97-6
Boiling Point 356.72°C
Atomic Weight 200.59
Quantity 1 lb.
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 0.002 mmHg at 25°C
Form Liquid
Molecular Formula Hg
MDL Number MFCD00011035
Synonym hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
SMILES [Hg]
IUPAC Name mercury
Molecular Weight (g/mol) 200.59
PubChem CID 23931
ChEBI CHEBI:16170
Formula Weight 200.59
Grade Certified ACS
Chemical Name or Material Mercury
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.