Learn More
Perkin Elmer US LLC Mercury (Hg) System 2 Electrodeless Discharge Lamp
Catalog No. NC1784242
Encompass_Preferred
System 2 Electrodeless Discharge Lamp for the detection of elemental Mercury (Hg). Note: PinAAcle™ instruments require the purchase of an EDL driver assembly, part number 03030997, for each of the lamp positions to be used. Electrodeless Discharge Lamps for greater brightness (EDLs) are typically much brighter and, in some cases, provide better sensitivity than comparable HCLs. EDLs are preferred for certain volatile elements. They offer better precision and lower detection limits for analyses that are “noisy” due to weak hollow cathode emission. Electrodeless Discharge Lamps (EDLs) are designed to be physically interchangeable with Intensitron Hollow Cathode Lamps. Their power supplies are built into all the PinAAcle™ series of atomic absorption spectrometers. Use of EDLs with those instruments requires only the purchase of an EDL driver assembly for each of the lamp positions to be used.
Catalog No. NC1784242 Supplier Perkin Elmer US LLC Supplier No. N3050634
System 2 Electrodeless Discharge Lamp for the detection of elemental Mercury (Hg). Note: PinAAcle™ instruments require the purchase of an EDL driver assembly, part number 03030997, for each of the lamp positions to be used. Electrodeless Discharge Lamps for greater brightness (EDLs) are typically much brighter and, in some cases, provide better sensitivity than comparable HCLs. EDLs are preferred for certain volatile elements. They offer better precision and lower detection limits for analyses that are “noisy” due to weak hollow cathode emission. Electrodeless Discharge Lamps (EDLs) are designed to be physically interchangeable with Intensitron Hollow Cathode Lamps. Their power supplies are built into all the PinAAcle™ series of atomic absorption spectrometers. Use of EDLs with those instruments requires only the purchase of an EDL driver assembly for each of the lamp positions to be used.