Learn More
Strem, An Ascensus Company CAS# 1600-27-7. 25g. Mercury(II) acetate, 98%. MFCD00012165
SDP
Catalog No. 5090116432
Encompass
CAS# 1600-27-7. 25g. Mercury(II) acetate, 98%. MFCD00012165. Molecular Weight: 318.68. Molecular Formula: Hg(OOCCH3)2. Color/form: white to light yellow pwdr. Strem# 93-8040. http://www.strem.com/catalog/v/93-8040/
Catalog No. 50-901-16432 Supplier Strem, An Ascensus Company Supplier No. 93804025G
CAS# 1600-27-7. 25g. Mercury(II) acetate, 98%. MFCD00012165. Molecular Weight: 318.68. Molecular Formula: Hg(OOCCH3)2. Color/form: white to light yellow pwdr. Strem# 93-8040. http://www.strem.com/catalog/v/93-8040/