Learn More
Mercury(II) acetate, ACS, 98.0% min
Description
Mercuric acetate is used as an oxidizing agent in organic synthesis. It is used in oxymercuration of double bonds. Mercuric acetate is used in non-aqueous titration. It is employed in the manufacture of phenyl mercury compounds which have pharmaceutical applications. It removes the acetamidomethyl protecting group from protected thiol, and converts thiocarbonate esters into dithiocarbonates. It promotes the addition of hydroxide and alkoxide across carbon-carbon double bonds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 179°C to 182°C |
| Color | Red |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Odor | Weak |
| Sensitivity | Light sensitive |
| Appearance | White crystals |
| Assay Percent Range | 98.0% min. |
| Density | 3.28 |
| Beilstein | 3563831 |
| Show More |
Safety and Handling
| Hazard Category | H300+H310+H330-H336-H373 |
| Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: II |
| EINECSNumber | 216-491-1 |
| RTECSNumber | AI8575000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.